Skip to Content
Merck
All Photos(1)

Key Documents

CLS3765

Corning® 384 well Microplates

flat bottom clear, white polystyrene, Tissue Culture (TC)-treated surface, bag of 25 ea, lid, sterile

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
R$5,767.00

R$5,767.00


Check Cart for Availability


Select a Size

Change View
100 EA
R$5,767.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear bottom
flat bottom clear
white polystyrene
size:
384 wells

R$5,767.00


Check Cart for Availability

material

clear bottom
flat bottom clear
white polystyrene

sterility

sterile

feature

lid
skirt

packaging

case of 100 ea
bag of 25 ea

manufacturer/tradename

Corning 3765

size

384 wells

well size

384

well volume

112 μL

wells

384 wells

working volume

20-80 μL , low flange

suitability

suitable for tissue

binding type

Tissue Culture (TC)-treated surface

General description

White walled plates enhance luminescent signals and have low background luminescence and fluorescence Opaque walls to prevent well-to-well crosstalk Optically clear flat well bottom permits direct microscopic viewing Can be used for both top and bottom reading instruments Flat bottoms with 112 μL total volume Recommended working volumes of 20 to 80 μL TC-treated for optimal cell attachment Sterilized by gamma radiation and certified nonpyrogenic Individual alphanumeric codes for well identification

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service