Skip to Content
Merck
All Photos(1)

Documents

CLS3628

Corning® 96 Well Clear Polystyrene Microplate

flat bottom clear, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.13

material

flat bottom clear
flat bottom polystyrene
flat wells

sterility

sterile

feature

lid
skirt

packaging

bag of 20

manufacturer/tradename

Corning 3628

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

clear

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® 96 well clear polystyrene microplate is sterile and nonpyrogenic. It is used for cell culture applications.

Features and Benefits

  • Corning® 96 well clear microplates support various application requirements.
  • Tissue culture-treated (TC-treated) polystyrene surface is used for the attachment and growth of anchorage-dependent cells.
  • The flat bottom is usually the best choice for detection.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alba Chavez-Dozal et al.
Journal of bacteriology, 203(3) (2020-11-18)
Vibrio fischeri is a cosmopolitan marine bacterium that oftentimes displays different colony morphologies, switching from a smooth to a wrinkly phenotype in order to adapt to changes in the environment. This wrinkly phenotype has also been associated with increased biofilm
Abdisalan M Noor et al.
BMC infectious diseases, 11, 121-121 (2011-05-17)
Countries aiming for malaria elimination require a detailed understanding of the current intensity of malaria transmission within their national borders. National household sample surveys are now being used to define infection prevalence but these are less efficient in areas of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service