CLS3564
Corning® disposable aseptic transfer caps for GL45 plastic spinner flasks
aseptic transfer cap for 3 L plastic spinner flasks, 1/8" dip tube, 0.2 μm vent, male luer lock, sterile, 2/cs
Synonym(s):
Corning cap accessory
About This Item
Recommended Products
material
polyethylene cap (filling cap)
polystyrene
sterility
sterile
feature
disposable
thread finish: 45 GL
packaging
pack of 1
case of 2
manufacturer/tradename
Corning 3564
color
orange cap
Related Categories
General description
- Cap is made of high density polyethylene; autoclaving is not recommended.
- The venting port ends in a 25 mm diameter vent filter with a 0.2 μm pore PVDF membrane and a barbed fitting.
- The liquid transfer port has an internal end with a rigid polypropylene dip tube angled to reach the bottom of the flask and on the external side a 1/8 in. I.D. (3.2 mm), 1/4 in. O.D. C-Flex® tubing ending in a male Luer lock (MLL) connector closed with a female Luer sealing cap.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service