Skip to Content
Merck
All Photos(1)

Key Documents

CLS3552

Corning® 96 well protein crystallization plates

COC, non-treated, 3:1 2uL round bottom wells, clear, nonsterile

Synonym(s):

96 well protein crystallization plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear plate
cyclic olefin copolymer (COC)
round bottom clear wells
size:
96 wells
Pricing and availability is not currently available.

material

clear plate
cyclic olefin copolymer (COC)
round bottom clear wells

sterility

non-sterile

feature

lid: no
plate format: 96 well
protein wells: 3/reservoir well

packaging

pack of 10 ea
case of 50 ea

manufacturer/tradename

Corning 3552

reservoir volume

100 μL

size

96 wells

well size

2 μL , protein well volume

well volume

100 μL

color

clear

suitability

suitable for (sitting drop vapor diffusion high-throughput (HT) protein crystallization)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Each well contains three 2μL round bottom protein wells and one 100μL reagent reservoir
  • Low volume 100μL reagent reservoir saves on reagent costs
  • Designed for sitting drop vapor diffusion high throughput protein crystallization
  • Manufactured from an advanced Cyclic Olefin Copolymer (COC) with high resistance to commonly used solvents, including acetone, acetic acid, butanone, ethanol, isopropanol, methanol, DMSO, nitric acid (65%), sulfuric acid (40%), hydrochloric acid (36%), and ammonia solution (33%) and improved transparency
  • Allows protein crystals to be easily viewed under polarized light with minimal background interference
  • Low water absorption of the polymer prevents loss of protein drop volume
  • Reservoir numbers are embossed on each individual well for easy identification
The Corning 96 well protein crystallization plate features large reservoir wells with corresponding protein wells. It is made of an advanced optical polymer (cyclic olefin copolymer (COC)) superior to polypropylene and polystyrene, which has higher optical clarity and greater resistance to organic solvents. The plate can be used for the sitting-drop method for high-throughput protein crystallization, and is compatible with manual pipettors and robotic operations.

The large reservoir well has a maximum volume of 100 μL, with a recommended working volume between 25 to 755 μL for screen reagents. The small protein well has a volume range of 1-4 μL.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service