CLS354536
Corning® BioCoat™ Poly-D-Lysine Cell Culture Flask
capacity 25 cm2, rectangular flask, canted neck, vented, Cap
Synonym(s):
cell culture flask, plastic cell culture flask, coated flask
About This Item
Recommended Products
material
rectangular flask
feature
vented Cap
canted neck
packaging
pkg of 10 ea (1 pack of 10)
manufacturer/tradename
Corning 354536
capacity
25 cm2
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service