Skip to Content
Merck
All Photos(1)

Documents

CLS353492

Corning® Falcon® Permeable Support

for use with 24 Well Plate, with 3 µm Translucent PET Membrane, sterile

Synonym(s):

Permeable Support

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

sterility

sterile

feature

with 3 µm Translucent PET Membrane

packaging

case of 48 ea (Individually wrapped)

manufacturer/tradename

Corning 353492

compatibility

for use with 24 Well Plate

Looking for similar products? Visit Product Comparison Guide

General description

Falcon® Permeable Support for 24 Well Plate with 3.0µm Translucent High Density Polyester (PET) Membrane, Sterile, 1/Pack, 48/Case

Application

Cell Culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service