CLS3517
Corning® Netwells® accessories
black reagent tray
Synonym(s):
cell culture dishes
Select a Size
R$1,078.00
Select a Size
About This Item
R$1,078.00
Recommended Products
material
polystyrene
sterility
non-sterile
feature
lid: no
packaging
case of 200 ea
pack of 25 ea
manufacturer/tradename
Corning 3517
size
1 well
color
black
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service