Skip to Content
Merck
All Photos(1)

Documents

CLS3340

Corning® 96 Well CellBIND® Microplates

flat bottom clear, black polystyrene, sterile, lid

Synonym(s):

96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, 96 well tissue culture treated plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

black plate
black polystyrene
flat bottom clear
flat bottom wells
polystyrene plate
round wells

sterility

sterile
sterile

feature

lid
lid
skirt
plate format: 96 well standard

packaging

pack of 5
case of 50

manufacturer/tradename

Corning 3340

size

96 wells

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

well volume

360 μL

well working volume

75-200 μL

color

black

binding type

CellBIND® surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Flat bottoms with 360 μL total volume
  • Recommended working volumes of 75 to 200 μL
  • CellBIND® surface treated for optimal cell attachment
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service