Skip to Content
Merck
All Photos(1)

Key Documents

CLLS1011

Sigma-Aldrich

SW48 Cells BAX/BAK (-/-,-/-)

Human female colon, colorectal adenocarcinoma

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
R$17,686.00

R$17,686.00


Estimated to ship onMay 08, 2025


Request a Bulk Order

Select a Size

Change View
1 SET
R$17,686.00

About This Item

UNSPSC Code:
41106514

R$17,686.00


Estimated to ship onMay 08, 2025


Request a Bulk Order

Product Name

SW48 Cells BAX/BAK (-/-,-/-),

biological source

human female colon (Source Disease: Colorectal adenocarcinoma)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... BAK(578) , BAX(581)

General description

This product corresponds to ATCC Cat. No. CCL-231

Application

Media Renewal changes one to two times per week
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture Ratio: approx. 1:2 to 1:6
The base medium for this cell line is DMEM, Cat. No. D5671. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10%, Sodium pyruvate, Cat. No. S8636, at a final concentration of 1 mM, and L-glutamine, Cat. No. G7513, at a final concentration of 2mM.
The cell freezing medium is DMSO 1X (Cat. No. C6164)

Features and Benefits

These SW48 cells are adherent, with a doubling time of approx. 35 hours.
Zinc finger nuclease (ZFN) double knock out on chromosome 19q13.3-13.4 / 6p21.3

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • SW48 Cells BAX/BAK (-/-,-/-) 1 vial

  • SW48 Control Cells 1 vial

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Srividya B Balachander et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(24), 6535-6549 (2020-09-30)
Targeting Bcl-2 family members upregulated in multiple cancers has emerged as an important area of cancer therapeutics. While venetoclax, a Bcl-2-selective inhibitor, has had success in the clinic, another family member, Bcl-xL, has also emerged as an important target and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service