Skip to Content
Merck
All Photos(8)

Key Documents

CLL1149

Sigma-Aldrich

A549 Cells GFP-CTNNB1 RFP-LMNB1

human male lung (Source Disease: Lung carcinoma)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
R$17,686.00

R$17,686.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
R$17,686.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

R$17,686.00


Check Cart for Availability

Request a Bulk Order

Product Name

A549 Cells GFP-CTNNB1 RFP-LMNB1,

biological source

human male lung (Source Disease: Lung carcinoma)

OMIM accession no.

storage temp.

−196°C

Gene Information

General description

A549 GFP-CTNNB1 RFP-LMNB1 are lung carcinoma cells from a human 58 year old caucasian male having two distinct ZFN modifications creating two fluorescent transgenes each expressed from their endogenous gene locus.

This cell line was derived from ATCC Catalog No. CCL-185.

Application

This product is a human A549 cell line in which the genomic CTNNB1 gene has been endogenously tagged with the 3XFLAG epitope linked to a Green Fluorescent Protein (GFP) gene and the LMNB1 gene has been endogenously tagged with a Red Fluorescent Protein (RFP) gene using CompoZr® zinc finger nuclease (ZFN) technology. Integration resulted in endogenous expression of the fusion proteins in which 3XFLAG-GFP is attached to the N-terminus of CTNNB1 and RFP is attached to the N-terminus of LMNB1. Fluorescence imaging shows CTNNB1 expression throughout the cell and LMNB1 expression localized to the nucleus. Upon activation with LiCl/leptomycin B or the Wnt3a ligand, the proteolysis of CTNNB1 is reduced so that translocation into the nucleus can be observed, making the cell line useful for high content screening to identify compounds that modulate this redistribution. The stable cell line was expanded from a single cell clone. CTNNB1 and LMNB1 gene regulation and corresponding protein function are preserved in contrast to cell lines exhibiting overexpression from an exogenous promoter.
To learn more, please visit the Cellular Reporter Cell Line webpage

Biochem/physiol Actions

β-catenin encoded by CTNNB1 gene is a multifunctional protein. It facilitates cell-cell adhesion differentiation and epithelial membrane transition (EMT). In addition, it plays a vital role in the WNT signaling pathway. Mutation in the gene leads to colorectal carcinomas.[1] Nuclear lamin B1 encoded by LMNB1 gene is an intermediate filament protein of the nuclear envelope.[2] The protein exhibits transcriptional co-regulatory activity and plays a key role in DNA replication, cellular aging and stress responses.[3] Overexpression of the gene has been observed in hepatocellular carcinoma (HCC) patients. Thus, LMNB1 can be used as a potential biomarker for HCC.[3]

Features and Benefits

Zinc Finger Nuclease (ZFN)-mediated targeted integrations of a fluorescent 3XFLAG-GFP just behind the ATG start codon of the CTNNB1 gene on chromosome 3p22.1 and a fluorescent RFP tag just behind the ATG start codon of the LMNB1 gene on chromosome 5q23.2 to create a cell line exhibiting stable expression of the two transgenes.

The A549 cells are adherent, with a doubling time of approx. 22 hours.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.

Preparation Note

Media Renewal changes two to three times per week.

Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.

Subculture Ratio: approx. 1:3-1:8

The base medium for this cell line is F12 Ham Medium, Cat. No. N4888. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10% and L-glutamine, Cat. No. G7513, at a final concentration of 2 mM.

Cell freezing medium-DMSO 1X, Cat. No. C6164.

Legal Information

3xFLAG is a trademark of Sigma-Aldrich Co. LLC
CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prognostic significance of COX-2 and ?-catenin in colorectal carcinoma.
Kazem A, et al.
Alexandria Journal of Medicine, 50(3), 211-220 (2014)
Structural Organization of the Human Gene (LMNB1) Encoding Nuclear Lamin B1
Lin F, et al.
Genomics, 27(2), 230-236 (1995)
Circulating Lamin B1 (LMNB1) Biomarker Detects Early Stages of Liver Cancer in Patients
Sun S, et al.
Journal of Proteome Research, 9(1), 70-78 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service