Skip to Content
Merck
All Photos(1)

Key Documents

CHA00011A

EUR-2019-nCoV E for detection of SARS-CoV-2 RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
NACRES:
NA.51
Pricing and availability is not currently available.

form

solution (in 10 mM Tris, 1 mM EDTA)

packaging

pkg of 1 vial

pH range

7.5-8.5

shipped in

ambient

storage temp.

−20°C

General description

EUR-2019-nCoV E Individual Assay is for qPCR (5′ nuclease assay) detection of SARS-CoV-2.

The product is primers and probes, the sequences of which were designed and made publicly available by Charité Institute of Virology – Universitätsmedizin Berlin.

For additional details, please visit SigmaAldrich.com/ncov.

Features and Benefits

The forward and reverse primers in each tube come with the following features:

  • DNA bases
  • No modifications
  • Desalt purified
  • 8 nmol each


The Dual-Labeled Probe in each tube comes with the following features:

  • DNA bases
  • 6-FAM reporter
  • BXQ (Block All Quencher—SigmaAldrich.com/bxq)
  • HPLC purified
  • 4nmol

Components

The product includes 1 individual tube of a 20X primer and probe pool for quantification of a SARS-CoV-2 gene.

Assay included is E_Sarbeco.The forward primer, reverse primer, and probe are at final concentrations of 8, 8, and 4 μM, respectively.

E_Sarbeco Assay:
Forward Primer ACAGGTACGTTAATAGTTAATAGCGT
Reverse Primer ATATTGCAGCAGTACGCACACA
Dual-Labeled Probe ACACTAGCCATCCTTACTGCGCTTCG

Legal Information

FAM is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service