Skip to Content
Merck
All Photos(1)

Key Documents

LB185

98062318, human blood, Lymphoblastoid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LB185, 98062318

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lymphoblastoid, normal,EBV-transformed

Cell Line Description

The lymphocyte cell line LB185 has been established using mononuclear cells from peripheral blood of a 56 year old normal male. T cells from the same sample have been used as controls for cellular radiosensitivity studies. Cells have been subsequently transformed with EBV. Fibroblasts have been derived from the same individual and were designated 153BR (Sigma Catalogue number. 90011808). This has been confirmed by STR profiling at ECACC. Cells are lymphhoblastoid with floating aggregates present.

Application

Mutation studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 11
D16S539: 9
D5S818: 10,13
D7S820: 11
THO1: 8,9
TPOX: 8,10
vWA: 17,18

Culture Medium

RPMI 1640 + 4mM Glutamine + 200 μg/ml Sodium Pyruvate (NaPy) + 15% Heat Inactivated (HI-FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml, dilute every 1-2 days to 3 x 100,000 cells/ml; 5% CO2, 37°C. Cells form floating aggregates.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service