Skip to Content
Merck
All Photos(1)

Documents

Nb2-11 Cell Line from rat

97041101, rat lymph, Lymphoblast

Synonym(s):

Nb2-11 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

Nb2-11 Cell Line from rat, 97041101, Thymus/lymph node, lymphoblast morphology

biological source

rat lymph

growth mode

Suspension

karyotype

42XY,-17,t(2,4)(q22,q24),t(10,11)(q12,q12),+der(15)t(15,?)(p12,?),-18,+der(18)t(18,?)(p12,?)

morphology

Lymphoblast

products

Not specified

receptors

Modified prolactin receptor (the Nb2 form)

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Rat lymphoma

Cell Line Description

Nb2-11 is a clone of the Nb-2 rat lymphoma line which was derived from a transplant of a lymphoma that developed in the thymus/lymph node of a male noble (Nb) strain rat following prolonged oestrogen treatment. The cells are of the pre-T cell origin and their proliferation is dependent on mammalian lactogens, such as prolactin. Nb2-11 can also be mitogenically stimulated by IL-2. Injection of Nb2 cells into Nb rats gives rise to malignant tumours that are highly sensitive to treatment with vinca alkaloids. Karyotypic analysis has shown that the cell line has only five well developed chromosome abnormalities. The cells do not express surface immunoglobulin, and their lactogen dependency was confirmed before deposit. Protocols for the use of Nb2-11 cells in bioassays are available from ECACC on request.

Application

1) in vitro bioassay of lactogenic hormones.
2) Studies of the mechanisms of lactogen induced mitogenesis.
3) Studies into the action of vinca alkaloids.
4) Studies of tumour progression.

Culture Medium

Fischer′s medium + 10% Fotal Bovine Serum (FBS) + 10% Horse Serum (HS) (gelding) + 0.075% Sodium Bicarbonate + 0.05 mM 2-Mercaptoethanol (2ME) + 2 mM Glutamine.

Subculture Routine

At resuscitation seed cultures at 3-5 x 100,000 cells/ml. Once established cultures should be seeded at 4 x 1,000 and 12 x 1,000 cells/ml for culture periods of 96 and 72 hours respectively. Do not exceed 9 x 100,000 cells/ml. Avoid using dilutions of cells with fresh medium at greater than 1:10 ratios; culture doubling time 12 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service