b.End5
96091930, mouse brain, Endothelial-like
Synonym(s):
Brain-derived Endothelial 5 Cells, bEnd3 Cells, bEnd5 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
b.End5, 96091930
biological source
mouse brain
growth mode
Adherent
karyotype
Not specified
morphology
Endothelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Related Categories
Cell Line Origin
Mouse Balb/c brain endothelioma
Cell Line Description
The cell line b.End5 has been established from brain endothelial cells of BALB/c mice. Immortalisation has been carried out by infection of primary cells with retrovirus coding for the Polyoma virus middle T-antigen. b.End5 cells are positive for endothelial specific proteins (PECAM-1, Endoglin, MECA-32, Flk-1) tested by FACS. Inflammatory cytokines induce the expression of proteins such as CAM-1, VCAM-1 and E-selectin. Injection into mice causes development of haemangiomas.
Application
Study of endothelial cell growth, differentiation and influence of oncogenes.
Culture Medium
DMEM + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 i.e. seeding at 4x10,000 cells/cm² using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service