Skip to Content
Merck
All Photos(1)

Documents

BF-11

Reactivity: Anti-Erythropoietin, human, 95050413

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Mouse Hybridoma

ImmunoDon

Rat AC1 spleen cells

Myeloma

Sp2/0 Ag-14

Reactivity

Anti-Erythropoietin, human

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat ACI spleen x Mouse Sp2/0 Ag-14

Cell Line Description

The BF-11 hybridoma cell line secretes a rat monoclonal antibody named Epoclonalan that reacts with human erythropoietin. Spleen cells from ACI strain rats that had been immunised with human urinary erythropoietin have been fused with Sp2/0 Ag-14 to produce this cell line. The α, β and asialo forms of human erythropoietin are bound by the antibody to the same extent.

Immunogen

Human erythropoietin

Application

Immunological studies, isolation of erythropoietin by immuno-affinity methods.
The isotype of the monoclonal antibody produced by this cell line is rat IgG2a.

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 100,000 - 1,000,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service