A2780ADR Cell Line human
93112520, human ovary, Epithelial
Synonym(s):
A2780-ADR Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
A2780ADR Cell Line human, 93112520
biological source
human ovary
description
Human Caucasian ovarian carcinoma
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian ovarian carcinoma
Cell Line Description
The adriamycin-resistant cell line A2780ADR has been developed by exposure of the parent A2780 cell line (Sigma catalogue no. 93112519) to adriamycin. A2780ADR is cross-resistant to melphalan and vinblastine. To retain resistance adriamycin has to be added to the media. The cells grow as a monolayer and in suspension in spinner cultures and are tumourigenic in immune deficient mice. Together with the cisplatin-resistant variant A2780cis these lines only differ in their exposure to a single drug and should facilitate the search for molecular changes responsible for the expression of pleiotropic drug resistance in human ovarian cancer.
Application
Study of drug transport, toxicity testing, drug resistance cancer studies
DNA Profile
STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 13
D16S539: 11,14
D5S818: 11,12
D7S820: 10
THO1: 6
TPOX: 8,9
vWA: 15,16
CSF1PO: 10,11
D13S317: 13
D16S539: 11,14
D5S818: 11,12
D7S820: 10
THO1: 6
TPOX: 8,9
vWA: 15,16
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS); treatment with 10E-7 M adriamycin at least once a week.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 5x1,000 to 2x10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C. Recommendation: culture cells without drug after resuscitation until growth has been fully established.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service