Skip to Content
Merck
All Photos(1)

Key Documents

COLO 677

93072109, human blood, Round & Refractile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

COLO 677, 93072109

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Round & Refractile

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human multiple myeloma

Cell Line Description

COLO 677 was originally described as being derived from a tumour in the left axillary lymph node of a 39-year-old male with small lung cell carcinoma in 1989. However, DNA fingerprinting suggests cross-contamination with cell line RPMI-8226; RPMI-8226 was established from the peripheral blood of a 61-year-old male with multiple myeloma in 1966 (see DSMZ website)

Application

Tumourigenicity studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Will form clusters in suspension.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service