Skip to Content
Merck
All Photos(1)

Key Documents

S2.2

Reactivity: Anti-Swine Testis (ST) cell antigen, 92040117

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Pig x Pig Hybridoma

packaging

tube of 5 μg 92040117-DNA-5UG
pkg of vial of cells 92040117-1VL

ImmunoDon

Boar spleen (d/d haplotype)

Myeloma

L231 (lymphoblastoid)

Reactivity

Anti-Swine Testis (ST) cell antigen

technique(s)

cell culture | mammalian: suitable

isotype

IgA

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Pig L231 x boar spleen (d/d haplotype)

Cell Line Description

The antibody is directed against swine testis cell antigen as determined in virus enzyme linked cell immunoassays (VELCIA). The antibody does not cross-react with mouse 3T3 cells.

Immunogen

Swine testis cell culture supernatant containing transmissible gastro-enteritis virus (strain Nouzilly 188.SG)

Application

VELICA, ELISA

Culture Medium

RPMI 1640 + 2mM glutamine + 1mM Sodium Pyruvate (NaP) + 0.05mM 2-mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 2-10x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service