GAD-1
Reactivity: Anti-Glutamic acid decarboxylase, 92020601
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
Mouse x Mouse Hybridoma
ImmunoDon
BALB/c mouse spleen
Myeloma
Sp2/0
Reactivity
Anti-Glutamic acid decarboxylase
technique(s)
cell culture | mammalian: suitable
isotype
IgG1
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse Sp2/0 x Mouse BALB/c spleen
Cell Line Description
The antibody is directed against glutamic acid decarboxylase from chickens, mice, rats, rabbits, cats and macaque monkeys.
Immunogen
Glutamic acid decarboxylase
Culture Medium
DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 2-6x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service