Skip to Content
Merck
All Photos(1)

Documents

AV3 (HeLa derivative)

88102402, human cervix, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

AV3 (HeLa derivative), 88102402

biological source

human cervix

growth mode

Adherent

karyotype

2n = 46

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma

Cell Line Description

This cell line was originally derived from normal human amnion. The cells are mainly used for virus studies (susceptible to poliovirus 1). HeLa marker chromosomes and type A G6PD have been detected. This cell line was found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line must be considered to be derived from HeLa.

Application

Virus studies: poliovirus 1

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,10
D13S317: 14
D16S539: 9,10
D5S818: 11,12
D7S820: 12
THO1: 7
TPOX: 8,12
vWA: 16,18

Culture Medium

EMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-5x100,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service