Skip to Content
Merck
All Photos(1)

Documents

HS-Sultan

87012701, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HS-Sultan, 87012701

biological source

human blood

growth mode

Suspension

karyotype

Hyperdiploid modal no=48

morphology

Lymphoblast

products

Immunoglobulin G (IgG) kappa chains in spinner culture. Endoplasmic reticulum in stationary culture.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Burkitt′s Lymphoma

Cell Line Description

It has been clearly established that HS-Sultan is a subclone of the Burkitt lymphoma derived cell line JIYOYE and not a myeloma cell line.

DNA Profile

STR-PCR Data:
Amelogenin: X,Y
CSF1PO: 10,11
D13S317: 12
D16S539: 10,11
D5S818: 12
D7S820: 8,10
THO1: 7,9
TPOX: 6,8
vWA: 15,19

Culture Medium

RPMI 1640 + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C. A density of 3x1,000,000 cells/ml may be achieved.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service