Skip to Content
Merck
All Photos(1)

Key Documents

MRC-9

85020202, human lung (fetal), Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

MRC-9, 85020202

biological source

human lung (fetal)

growth mode

Adherent

karyotype

2n = 46, diploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian foetal lung

Cell Line Description

Established from normal lung tissue of a 15 week old female Caucasian foetus. Like MRC-5 the line is susceptible to a wide range of viruses. At least 40 population doublings expected.

Application

Virus studies: rhinoviruses, production of viral vaccines

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 8,11
D16S539: 12
D5S818: 12,13
D7S820: 10,11
THO1: 6,8
TPOX: 11
vWA: 16,17

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service