Skip to Content
Merck
All Photos(1)

Key Documents

108CC5-TG-3

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Rat Hybridoma nerve

packaging

tube of 5 μg 08062545-DNA-5UG
pkg of vial of cells 08062545-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma x Rat glioma hybrid

Cell Line Description

A 6-thioguanine resistant mutant of 108CC5 (Sigma Catalogue number 08062521), a mouse neuroblastoma N18TG2 x rat glioma C6-BU-1 hybrid

Application

Used as a neuronal Model

Culture Medium

DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + 0.1 mM 6-thioguanine. The cells can also be grown in the absence of 6-thioguanine.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-2x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C. The cultures rapidly produce lactic acid through anaerobic glycolysis causing the media to become acidified. The medium must never be allowed to become acidic as cells detach in clumps and take many passages to recover. Frequent inspection of cultures is therefore required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service