Skip to Content
Merck
All Photos(1)

Key Documents

CASP8C

Sigma-Aldrich

Caspase 8 Assay Kit, Colorimetric

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.84

shipped in

dry ice

Quality Level

storage temp.

−20°C

Gene Information

human ... CASP8(841)

General description

Caspases (cysteine-requiring aspartate proteases) belong to a highly conserved family of cysteine proteases with specificity for aspartic acid residues found in their substrates. Caspases play a central role in apoptosis. Inhibition of caspases prevents the cell death process. Currently, there are 13 caspases recognized in mammals. Caspases are classified as initiators and effectors. Caspase 8 belongs to the class of initiators. It possesses a prodomain at the terminal and contains the death effector domain which serves as protein-protein interaction motif for caspase 8. With respect to substrate specificity, caspase 8 is classified as a member of group III. It specifically recognizes the (I/L/V) EXD sequence on the substrate.

Application

The Caspase 8 Colorimetric Assay Kit is based on the hydrolysis of the peptide substrate Acetyl-Ile-Glu-Thr-Asp-p-Nitroaniline (Ac-IETD-pNA) by caspase-8 that results in the release of a p-Nitroaniline (p-NA) moiety. The p-NA is read at 405 nm. The concentration of the p-NA released can be calculated from a calibration curve prepared from p-NA standards (included with the kit).
The kit detects caspase 8 activity in crude and purified preparations of caspase 8.
Complete kit contains:
  • purified caspase 8 for a positive control
  • caspase 8 inhibitor
  • p-NA fluorescent standard

Quantity

The kit provides sufficient reagents for 100 tests of 100 μL in 96-well plates.

Kit Components Only

Product No.
Description

  • N-Acetyl-Ile-Glu-Thr-Asp-al, ≥97% (HPLC) 100 μL

  • Acetyl-Ile-Glu-Thr-Asp-p-nitroaniline Substrate 100 μL

  • 10X Assay Buffer 5 mL

  • 5X Lysis Buffer 5 mL

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RNAi-Mediated Simultaneous Downregulation of uPAR and Cathepsin B Induces Caspase 8-Mediated Apoptosis in SNB19 Human Glioma Cells
Gondi CS, et al.
Molecular Cancer Therapeutics, 5(12), 3197-3197 (2006)
Graeme W Carlile et al.
FEBS letters, 583(4), 848-854 (2009-02-27)
Issues remain to be elucidated in the developmental regulation of erythropoiesis. In particular the role of Fas, a member of the tumor necrosis factor family of receptors despite much work remains unclear. During erythropoiesis, Fas is expressed at low levels
Basic Concepts of Molecular Pathology, 32-32 (2009)
Parisa Habibi et al.
Iranian journal of basic medical sciences, 23(1), 79-85 (2020-05-15)
The useful and effective role of exercise program to prevent cardiac tissue apoptosis and fibrosis in ovariectomized type 2 diabetic (T2DM) rats (OVR.D) is well known. The current study aimed to investigate the simultaneous effects of T2DM and swimming plan
Febrile-Range Hyperthermia Accelerates Caspase-Dependent Apoptosis in Human Neutrophils
Nagarsekar A, et al.
Journal of Immunology, 181, 2638-2638 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service