Skip to Content
Merck
All Photos(1)

Key Documents

CAS9RFPVP

Sigma-Aldrich

Cas9-RFP Lenti Plasmid

Sign Into View Organizational & Contract Pricing

Select a Size

1 μG
R$2,096.00

R$2,096.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 μG
R$2,096.00

About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

R$2,096.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

form

liquid

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of Plasmid DNA)

application(s)

CRISPR

reporter gene

RFP

selection

ampicillin

shipped in

dry ice

storage temp.

−20°C

General description

This product is a lentiviral plasmid that utilizes the EF1 alpha promoter to drive expression of Cas9 and RFP linked by a 2A peptide (EF1a-Cas9-2A-RFP) allowing for easy visualization of successful transfection or transduction. Use Sigma′s lentiviral Cas9 plasmid for generation of lentiviral particles and efficient production of stable cell lines expressing Cas9 for CRISPR based genome editing. Sigma′s Cas9-RFP lenti plasmid is one part of a two part CRISPR system with individual Cas9 and gRNA expression vectors. For easy fluorescence microscopy a MOI greater than 200 should be employed when packaged into lentiviral particles.

To order gRNA in any format click here

Application

Functional Genomics/Target Validation
  • Creation of gene knockouts in multiple cell lines
  • Complete knockout of genes not amenable to RNAi
  • Manufacture of Cas9-RFP expressing Lentiviral Particles

Features and Benefits

  • Highly specific
  • Highly active
  • Ready to use purified plasmid DNA

Principle

CRISPR/Cas systems are employed by bacteria and archaea as a defense against invading viruses and plasmids. Recently, the type II CRISPR/Cas system from the bacterium Streptococcus pyogenes has been engineered to function in eukaryotic systems using two molecular components: a single Cas9 protein and a non-coding guide RNA (gRNA). The Cas9 endonuclease can be programmed with a single gRNA, directing a DNA double-strand break (DSB) at a desired genomic location. Similar to DSBs induced by zinc finger nucleases (ZFNs), the cell then activates endogenous DNA repair processes, either non-homologous end joining (NHEJ) or homology-directed repair (HDR), to heal the targeted DSB.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service