Skip to Content
Merck
All Photos(1)

Key Documents

C9913

Sigma-Aldrich

Cholesterol

SyntheChol® synthetic, ≥98%

Synonym(s):

3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Beilstein:
1915888
EC Number:
MDL number:
UNSPSC Code:
51101500

biological source

synthetic

Assay

≥98%

bp

360 °C (lit.)

mp

147-149 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Preparation Note

Synthesized from material of non-animal origin

Legal Information

SyntheChol is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinjiang Fan et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(23), 7261-7266 (2015-06-04)
Translocator protein (TSPO) is a key member of the mitochondrial cholesterol transport complex in steroidogenic tissues. To assess the function of TSPO, we generated two lines of Cre-mediated Tspo conditional knockout (cKO) mice. First, gonadal somatic cell-targeting Amhr2-Cre mice were
Matthew P Buman et al.
American journal of epidemiology, 179(3), 323-334 (2013-12-10)
Sleep and sedentary and active behaviors are linked to cardiovascular disease risk biomarkers, and across a 24-hour day, increasing time in 1 behavior requires decreasing time in another. We explored associations of reallocating time to sleep, sedentary behavior, or active
Daniel I Swerdlow et al.
Lancet (London, England), 385(9965), 351-361 (2014-09-30)
Statins increase the risk of new-onset type 2 diabetes mellitus. We aimed to assess whether this increase in risk is a consequence of inhibition of 3-hydroxy-3-methylglutaryl-CoA reductase (HMGCR), the intended drug target. We used single nucleotide polymorphisms in the HMGCR
Lingyun Lu et al.
American heart journal, 167(5), 683-689 (2014-04-29)
We previously found that the use of ezetimibe increased rapidly with different patterns between the United States (US) and Canada prior to the landmark Ezetimibe and Simvastatin in Hypercholesterolemia Enhance Atherosclerosis Regression (ENHANCE) trial, which was reported in January 2008
Steven A Grover et al.
The lancet. Diabetes & endocrinology, 3(2), 114-122 (2014-12-09)
Despite the increased risk of cardiovascular disease and type 2 diabetes associated with excess bodyweight, development of a clinically meaningful metric for health professionals remains a challenge. We estimated the years of life lost and the life-years lost from diabetes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service