Skip to Content
Merck
All Photos(1)

Documents

C9187

Sigma-Aldrich

Cys-Gly, oxidized

≥95% (HPLC)

Synonym(s):

Cys(Cys-Gly)-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18N4O6S2
CAS Number:
Molecular Weight:
354.40
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥95% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

NC(CSSCC(N)C(=O)NCC(O)=O)C(=O)NCC(O)=O

InChI

1S/C10H18N4O6S2/c11-5(9(19)13-1-7(15)16)3-21-22-4-6(12)10(20)14-2-8(17)18/h5-6H,1-4,11-12H2,(H,13,19)(H,14,20)(H,15,16)(H,17,18)

InChI key

KDJVKDYFFTWHBO-UHFFFAOYSA-N

Biochem/physiol Actions

Cys(Cys-Gly)-Gly is the reversibly oxidized form of Cys-Gly. L-Cysteine-L-Glycine (Cys-Gly) is used in studies on the homeostasis of glutathione. Cys-Gly is an important molecule for study in the areas of thiol homeostasis, oxidative stress and metals management.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hardeep Kaur et al.
The Journal of biological chemistry, 284(21), 14493-14502 (2009-04-07)
GSH metabolism in yeast is carried out by the gamma-glutamyl cycle as well as by the DUG complex. One of the last steps in the gamma-glutamyl cycle is the cleavage of Cys-Gly by a peptidase to the constitutent amino acids.
Hardeep Kaur et al.
Biochimie, 93(2), 175-186 (2010-09-28)
Dug1p is a recently identified novel dipeptidase and plays an important role in glutathione (GSH) degradation. To understand the mechanism of its substrate recognition and specificity towards Cys-Gly dipeptides, we characterized the solution properties of Dug1p and studied the thermodynamics
Elena Chekmeneva et al.
Metallomics : integrated biometal science, 3(8), 838-846 (2011-06-21)
Isothermal Titration Calorimetry (ITC) was used to study the binding of Cd(2+) by phytochelatins ((γGlu-Cys)(n)-Gly, PC(n); n = 1-5) and their selected fragments (Cys, Cys-Gly and γGlu-Cys) in order to understand the influence of the chain length on the complex
Thomas Müller et al.
Movement disorders : official journal of the Movement Disorder Society, 26(3), 543-546 (2011-04-05)
Oxidative stress is influenced by the thiol homeostasis, which determines the redox milieu. One of its components is Cysteinyl-glycine (Cys-Gly) generation, as its metabolic precursor is the free radicals scavenging glutathione. Levodopa is under suspicion to promote oxidative stress via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service