Skip to Content
Merck
All Photos(1)

Key Documents

C5738

Sigma-Aldrich

Anti-Calcium Channel (β2 subunit) (Voltage Gated Ca2+ Channel) antibody produced in rabbit

~1 mg/mL, fractionated antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

western blot: 5-10 μg/mL using brain tissues lysate

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CACNB2(783)
mouse ... Cacnb2(12296)
rat ... Cacnb2(116600)

General description

CACNB2 (Voltage-dependent L-type calcium channel subunit β-2) is particularly expressed in cardiovascular tissue.

Specificity

Specifically recognizes a splice variant of β2 (68 kDa) from human, mouse and rat.

Immunogen

synthetic peptide derived from the rat β2 calcium channel subunit conjugated to KLH.

Application

Anti-Calcium Channel (β2 subunit) antibody produced in rabbit is suitable for western blot at a concentration of 5-10μg/mL using rat brain tissue lysate.

Biochem/physiol Actions

CACNB2 (Voltage-dependent L-type calcium channel subunit β-2) is a subunit of voltage-activated L-type calcium channelsl. CACNB2 regulates voltage-dependent calcium currents through direct interaction with α1 subunits. Loss of function mutation in CACNB2 is associated with familial cardiac death combined with short QT intervals and ST-segment elevation. It is down-regulated in humans with atrial fibrillation and miR-21 is suggested to be responsible for the down-regulation. Mutations in CACNB2 are linked to autism spectrum disorders.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana Barana et al.
Circulation. Arrhythmia and electrophysiology, 7(5), 861-868 (2014-08-12)
Atrial fibrillation is characterized by progressive atrial structural and electrical changes (atrial remodeling) that favor arrhythmia recurrence and maintenance. Reduction of L-type Ca(2+) current (I(Ca,L)) density is a hallmark of the electrical remodeling. Alterations in atrial microRNAs could contribute to
Giuseppe Ronzitti et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(32), 10603-10615 (2014-08-08)
α-Synuclein is thought to regulate neurotransmitter release through multiple interactions with presynaptic proteins, cytoskeletal elements, ion channels, and synaptic vesicles membrane. α-Synuclein is abundant in the presynaptic compartment, and its release from neurons and glia has been described as responsible
Yuxin Niu et al.
Circulation. Cardiovascular genetics, 3(6), 548-555 (2010-12-16)
Single-nucleotide polymorphisms (SNPs) within the regulatory β2 subunit of the voltage-gated calcium channel (CACNB2) may contribute to variable treatment response to antihypertensive drugs and adverse cardiovascular outcomes. SNPs in CACNB2 from 60 ethnically diverse individuals were identified and characterized. Three
Alexandra F S Breitenkamp et al.
PloS one, 9(4), e95579-e95579 (2014-04-23)
Autism Spectrum Disorders (ASD) are complex neurodevelopmental diseases clinically defined by dysfunction of social interaction. Dysregulation of cellular calcium homeostasis might be involved in ASD pathogenesis, and genes coding for the L-type calcium channel subunits CaV1.2 (CACNA1C) and CaVβ2 (CACNB2)
Charles Antzelevitch et al.
Circulation, 115(4), 442-449 (2007-01-17)
Cardiac ion channelopathies are responsible for an ever-increasing number and diversity of familial cardiac arrhythmia syndromes. We describe a new clinical entity that consists of an ST-segment elevation in the right precordial ECG leads, a shorter-than-normal QT interval, and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service