Skip to Content
Merck
All Photos(1)

Key Documents

C5460

Sigma-Aldrich

Hexadecylpyridinium chloride monohydrate

BioXtra, 99.0-102.0%

Synonym(s):

Cetylpyridinium chloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38ClN · H2O
CAS Number:
Molecular Weight:
358.00
ISBN-10:
MFCD00149977
Beilstein:
3578606
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

cationic

Quality Level

product line

BioXtra

Assay

99.0-102.0%

form

powder

mol wt

358.00 g/mol

technique(s)

electrophoresis: suitable

impurities

≤0.001% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

solubility

H2O: 0.5 M, clear, colorless to faintly yellow

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.[Cl-].CCCCCCCCCCCCCCCC[n+]1ccccc1

InChI

1S/C21H38N.ClH.H2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-22-20-17-15-18-21-22;;/h15,17-18,20-21H,2-14,16,19H2,1H3;1H;1H2/q+1;;/p-1

InChI key

NFCRBQADEGXVDL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Hexadecylpyridinium chloride monohydrate is a cationic surfactant.

Application

Hexadecylpyridinium chloride monohydrate has been used in a study to assess the decontamination process for the isolation of Mycobacterium avium subspecies Paratuberculosis from fecal samples using ESP broth cultures. It has also been used in a study to investigate the removal of emulsified food and mineral oils from wastewater.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Removal of emulsified food and mineral oils from wastewater using surfactant modified barley straw
Ibrahim, S., et al.
Bioresource Technology, 100, 6-6 (2009)
Sreekumari Rajeev et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 19(6), 702-704 (2007-11-14)
Based on the authors' laboratory experience indicating that increased bacterial contamination in Mycobacterium avium ssp. paratuberculosis (MAP) cultures may be because of the addition of brain heart infusion broth (BHI) during the decontamination process, this study was designed to examine
Kwang Sik Suh et al.
Cytotechnology, 66(6), 925-935 (2013-10-12)
Achyranthes bidentata (A. bidentata) Blume is a medicinal herb with the property of strengthening bones and muscles and ensuring proper downward flow of blood in terms of the therapeutic theory of traditional medicine. In the present study, the effect of
Emilie Laurin et al.
Journal of dairy science, 98(12), 8597-8609 (2015-10-20)
Mycobacterium avium ssp. paratuberculosis (MAP) can be shed in feces, milk, and colostrum. The goal of this study was to assess assays that detect MAP in these sample types, including effects of lactation stage or season. Understanding the performance of
Kazuko Ishiguro et al.
Journal of microbiological methods, 115, 22-26 (2015-05-20)
Bacterial metabolic activity is associated with the onset and progression mechanisms of oral biofilm-mediated disease; however, at present no method to monitor bacterial metabolism exists, especially for periodontopathic bacteria. Therefore, we aimed to establish a novel method for monitoring the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service