Key Documents
C4176
Sigma-Aldrich
Casting clamps
For single-gel casting assembly
About This Item
UNSPSC Code:
12352200
NACRES:
NB.51
material
Nylon
General description
Specifically designed for gel casting. Wide jaws provide uniform pressure on glass plate-gasket assembly. Single- and dual-gel casting clamps accommodate different thicknesses and are not interchangeable.
Set of three (3). One set (three clamps) is sufficient for 10 cm wide mini-gels; two sets (six clamps) are necessary for standard 16.5 cm wide gels.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Tatsuya Osaki et al.
Scientific reports, 7(1), 1897-1897 (2017-05-17)
Consistent cell preparation is a fundamental preliminary step for understanding complex cellular mechanisms in various cell-based research fields, including basic cell biology, cancer research, and tissue engineering. However, certain elusive factors, such as cellular de-differentiation and contamination with mycoplasma or