Skip to Content
Merck
All Photos(1)

Key Documents

C3330

Sigma-Aldrich

Carcinoembryonic Antigen Fragment 101-115

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C81H126N20O22
CAS Number:
Molecular Weight:
1731.99
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥95% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CO)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCCN)NC(=O)[C@@H]2CCCN2C(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H]3CCCN3C(=O)[C@H](Cc4ccc(O)cc4)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](Cc5ccccc5)NC(=O)[C@H](CCC(N)=O)NC(=O)CNC(=O)[C@@H](N)[C@@H](C)O)C(C)C)C(O)=O

InChI

1S/C81H126N20O22/c1-8-45(6)66(80(122)123)98-71(113)57(42-102)96-74(116)60-24-15-35-99(60)77(119)53(20-12-13-33-82)92-73(115)59-23-17-36-100(59)78(120)55(38-43(2)3)94-68(110)52(30-32-63(107)108)91-72(114)58-22-16-37-101(58)79(121)56(40-48-25-27-49(104)28-26-48)95-76(118)65(44(4)5)97-69(111)50(21-14-34-87-81(85)86)90-70(112)54(39-47-18-10-9-11-19-47)93-67(109)51(29-31-61(83)105)89-62(106)41-88-75(117)64(84)46(7)103/h9-11,18-19,25-28,43-46,50-60,64-66,102-104H,8,12-17,20-24,29-42,82,84H2,1-7H3,(H2,83,105)(H,88,117)(H,89,106)(H,90,112)(H,91,114)(H,92,115)(H,93,109)(H,94,110)(H,95,118)(H,96,116)(H,97,111)(H,98,113)(H,107,108)(H,122,123)(H4,85,86,87)/t45-,46+,50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,64-,65-,66-/m0/s1

InChI key

LMDZISZNTCXHDM-KEDROYBBSA-N

Gene Information

Amino Acid Sequence

Thr-Gly-Gln-Phe-Arg-Val-Tyr-Pro-Glu-Leu-Pro-Lys-Pro-Ser-Ile

Biochem/physiol Actions

Peptide with strong affinity for the 80 kDa surface protein on Kupffer cells, possibly involved in hepatic metastasis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Thomas et al.
Biochemical and biophysical research communications, 188(2), 671-677 (1992-11-09)
Clearance of carcinoembryonic antigen (CEA) from the circulation is by binding to Kupffer cells in the liver. We have shown that CEA binding to Kupffer cells occurs via a peptide sequence YPELPK representing amino acids 107-112 of the CEA sequence.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service