Skip to Content
Merck
All Photos(2)

Key Documents

C2237

Sigma-Aldrich

Cypermethrin

≥98% (HPLC)

Synonym(s):

α-Cyano-(3-phenoxy­phenyl)­methyl 3-(2,2-dichloro­vinyl)-2,2-dimethyl­cyclo­pro­pane­carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19Cl2NO3
CAS Number:
Molecular Weight:
416.30
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

storage temp.

−20°C

SMILES string

CC1(C)C(\C=C(\Cl)Cl)C1C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2

InChI

1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3

InChI key

KAATUXNTWXVJKI-UHFFFAOYSA-N

Biochem/physiol Actions

Type II pyrethrin. A potent inhibitor of calcineurin (protein phosphatase 2B).

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin A Elser et al.
Toxicological sciences : an official journal of the Society of Toxicology, 175(2), 182-196 (2020-03-20)
Prenatal exposure to cypermethrin is a risk factor for adverse neurodevelopmental outcomes in children. In addition, maternal psychological stress during pregnancy has significant effects on fetal neurodevelopment and may influence end-stage toxicity to offspring by altering maternal xenobiotic metabolism. As
George P Lahm et al.
Bioorganic & medicinal chemistry letters, 15(22), 4898-4906 (2005-09-17)
A novel class of anthranilic diamides has been discovered with exceptional insecticidal activity on a range of Lepidoptera. These compounds have been found to exhibit their action by release of intracellular Ca2+ stores mediated by the ryanodine receptor. The discovery
María Carla Cecere et al.
Journal of medical entomology, 50(2), 394-403 (2013-04-02)
The effectiveness of two doses of suspension concentrate (SC) pyrethroid insecticides in suppressing peridomestic populations of Triatoma infestans (Klug) was evaluated in 28 rural communities located in Santiago del Estero province, northwestern Argentina, including 388 houses and 1,516 identified sites.
Hassane Adakal et al.
Experimental & applied acarology, 59(4), 483-491 (2012-09-13)
Traditional systems account for 95 % of the livestock produced in Burkina Faso. Tick infestation hampers livestock productivity in this area. However, little information exists on tick-control practices used by livestock farmers. We interviewed 60 stockbreeders working in traditional farming systems
Valentina Sechi et al.
Environmental pollution (Barking, Essex : 1987), 184, 586-596 (2013-11-10)
A soil multi-species, SMS, experimental test system consisting of the natural microbial community, five collembolan species and a predatory mite along with either Enchytraeus crypticus or the earthworm Eisenia fetida were exposed to α-cypermethrin. A comparison of the performance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service