Skip to Content
Merck
All Photos(1)

Key Documents

C2125

Sigma-Aldrich

Methyl decanoate

Sigma Grade, ~99%

Synonym(s):

Capric acid methyl ester, Decanoic acid methyl ester, Methyl caprate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOCH3
CAS Number:
Molecular Weight:
186.29
Beilstein:
1759170
EC Number:
MDL number:
UNSPSC Code:
12352211

grade

Sigma Grade

Assay

~99%

refractive index

n20/D 1.425 (lit.)

bp

108 °C/10 mmHg
224 °C (lit.)

mp

−14-−11 °C (lit.)

density

0.871 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCC(=O)OC

InChI

1S/C11H22O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h3-10H2,1-2H3

InChI key

YRHYCMZPEVDGFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.9 °F - Pensky-Martens closed cup

Flash Point(C)

110.5 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service