Skip to Content
Merck
All Photos(3)

Documents

C1623

Sigma-Aldrich

Anti-CLIC2 (233-247) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CLIC2b, Anti-Chloride intracellular channel protein 2, Anti-XAP121

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~28 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLIC2(1193)

General description

Chloride intracellular channel protein 2 is a protein encoded by the CLIC2 gene in humans and belongs to chloride intracellular channel (CLIC) family. It is also referred as CLIC2b, MRXS32 and XAP121. CLIC2 protein is a member of the glutathione transferase class of proteins and they can exist as integral membrane-channel form or a soluble form. The gene is possibly associated with one of the many diseases linked to Xq28.

Immunogen

synthetic peptide corresponding to amino acids 233-247 of human CLIC2

Application

Anti-CLIC2 (233-247) antibody produced in rabbit is suitable for western blotting at a dilution of 1:500-1:2,000.

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

Chloride intracellular channel 2 (CLIC2) functions by regulating ryanodine receptor (RyR) intracellular Ca(2+) release channels. RyR proteins play a major role in the regulation of Ca(2+) signaling in many cells. CLIC2 protein also helps in maintaining normal cognitive function by interacting with RyRs in the brain. In healthy individuals it is studied that the CLIC2 protein can tolerate amino acid substitutions and can be fully functional.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyoko Takano et al.
Human molecular genetics, 21(20), 4497-4507 (2012-07-21)
Chloride intracellular channel 2 (CLIC2) protein is a member of the glutathione transferase class of proteins. Its' only known function is the regulation of ryanodine receptor (RyR) intracellular Ca(2+) release channels. These RyR proteins play a major role in the
N S Heiss et al.
Genomics, 45(1), 224-228 (1997-10-27)
A transcript map was previously constructed in the 1200-kb telomeric region of Xq28. One of the cDNAs, XAP121, displayed homology to a p64 bovine chloride channel and to a human chloride channel (p64CLCP, NCC27) at both the nucleotide and the
Brett A Cromer et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 63(Pt 11), 961-963 (2007-11-17)
The chloride intracellular channel (CLIC) family of proteins are unusual in that they can exist in either an integral membrane-channel form or a soluble form. Here, the expression, purification, crystallization and preliminary diffraction analysis of CLIC2, one of the least-studied
Shawn Witham et al.
Proteins, 79(8), 2444-2454 (2011-06-02)
Large-scale next generation resequencing of X chromosome genes identified a missense mutation in the CLIC2 gene on Xq28 in a male with X-linked intellectual disability (XLID) and not found in healthy individuals. At the same time, numerous nsSNPs (nonsynonomous SNP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service