Skip to Content
Merck
All Photos(1)

Key Documents

C-14043

Sigma-Aldrich

Normal Human Epidermal Melanocytes 2 (NHEM 2)

Adult, isolated in PMA-free Melanocyte Growth Medium M2, 1 million cells per pellet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human epidermis (adult)

packaging

pkg of 1,000,000 cells

morphology

(epidermal)

application(s)

cell analysis (DNA, RNA, and protein)

shipped in

dry ice

storage temp.

−20°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.

Cell Line Origin

Skin

Application

Primary Normal Human Epidermal Melanocytes (NHEM) are isolated from the epidermis of adult skin from different locations including the face, the breasts, the abdomen, and the thighs. Melanocytes are located in the stratum basale, but branch out between the keratinocytes in suprabasal layers. About 5-10 % of the cells in the epidermis are melanocytes. The main purpose of melanocytes is the production of melanin, the protein responsible for the pigmentation of the skin, eyes, and hair. Melanin protects the cells in the skin and in deeper layers from the hazardous effects of UV radiation. It is produced and stored in melanosomes, which are located close to the melanocyte membrane, but it can also be transferred to neighboring keratinocytes.Normal Human Epidermal Melanocytes 2 are isolated using the serum-free and PMA-free Melanocyte Growth Medium M2. Since PMA is a tumor promoting mitogen, it can interfere with experimental approaches. Therefore, we recommend using cells isolated in Melanocyte Growth Medium M2 and also using this medium for cultivation. Normal Human Epidermal Keratinocytes (NHEK) and Normal Human Dermal Fibroblasts (NHDF) from the same donor are available on request.

Components

PMA-free Melanocyte Growth Medium M2

Quality

Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.

Physical form

Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.

Other Notes

The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.

Legal Information

RNAlater is a registered trademark of Ambion, Inc.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Primary Human Epidermal Melanocytes and optimized growth media support in vitro cultivation.

Primary cells from PromoCell are available in pelleted form for research applications like gene expression analysis, and cell/protein characterization experiments.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service