Skip to Content
Merck
All Photos(1)

Documents

C134

Supelco

Chlorpromazine hydrochloride

Synonym(s):

2-Chloro-10-(3-dimethylaminopropyl)phenothiazine hydrochloride, CPZ, Largactil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19ClN2S · HCl
CAS Number:
Molecular Weight:
355.33
Beilstein:
3779989
EC Number:
MDL number:
UNSPSC Code:
41116107

format

neat

SMILES string

Cl[H].CN(C)CCCN1c2ccccc2Sc3ccc(Cl)cc13

InChI

1S/C17H19ClN2S.ClH/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H

InChI key

FBSMERQALIEGJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Substitute for benzidine, o-dianisidine, and o-tolidine in the determination of microquantities of hemoglobin and peroxidase.

Biochem/physiol Actions

Chlorpromazine demonstrates cytotoxic and antiproliferative activity against leukemic cells, but does not affect the viability of normal lymphocytes.
Phenothiazine antipsychotic; D2 dopamine receptor antagonist, H1 histamine receptor antagonist; inhibits calmodulin-dependent stimulation of cyclic nucleotide phosphodiesterase and nitric oxide synthase.

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafaa Zeineddine et al.
Molecular neurodegeneration, 10, 57-57 (2015-11-02)
Amyotrophic Lateral Sclerosis is characterized by a focal onset of symptoms followed by a progressive spread of pathology that has been likened to transmission of infectious prions. Cell-to-cell transmission of SOD1 protein aggregates is dependent on fluid-phase endocytosis pathways, although
Jessica J W Broeders et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(3), 1057-1064 (2013-02-05)
The extrapolation of in vitro to in vivo toxicity data is a challenge. Differences in sensitivity between cell systems may be due to intrinsic properties of the cell but also because of differences in exposure. In this study, the cytotoxicity
Jessica M D'Amico et al.
Journal of neurophysiology, 109(6), 1473-1484 (2012-12-12)
In animals, the recovery of motoneuron excitability in the months following a complete spinal cord injury is mediated, in part, by increases in constitutive serotonin (5-HT2) and norepinephrine (α1) receptor activity, which facilitates the reactivation of calcium-mediated persistent inward currents
Soon Young Shin et al.
Carcinogenesis, 34(9), 2080-2089 (2013-05-22)
2-Chloro-10-[3(-dimethylamino)propyl]phenothiazine mono hydrochloride (chlorpromazine; CPZ) is an antipsychotic agent that was originally developed to control psychotic disorders. The cytotoxic properties of the CPZ are well known, but its mechanism of action is poorly understood. In this study, we investigated the
Stephanie L MacAllister et al.
Canadian journal of physiology and pharmacology, 91(1), 56-63 (2013-02-02)
Chlorpromazine (CPZ), a member of the largest class of first-generation antipsychotic agents, is known to cause hepatotoxicity in the form of cholestasis and hepatocellular necrosis in some patients. The mechanism of CPZ hepatotoxicity is unclear, but is thought to result

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service