Skip to Content
Merck
All Photos(1)

Key Documents

C1306

Sigma-Aldrich

BQ-123

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H41N6O7 Na
CAS Number:
Molecular Weight:
632.68
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

[H][C@@]12CCCN1C(=O)[C@@H](CC(O)=O)NC(=O)[C@@H](Cc3c[nH]c4ccccc34)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](NC2=O)C(C)C

InChI

1S/C31H42N6O7/c1-16(2)12-21-27(40)33-22(13-18-15-32-20-9-6-5-8-19(18)20)28(41)35-23(14-25(38)39)31(44)37-11-7-10-24(37)29(42)36-26(17(3)4)30(43)34-21/h5-6,8-9,15-17,21-24,26,32H,7,10-14H2,1-4H3,(H,33,40)(H,34,43)(H,35,41)(H,36,42)(H,38,39)/t21-,22+,23+,24-,26+/m0/s1

InChI key

VYCMAAOURFJIHD-PJNXIOHISA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

cyclo-Asp-Pro-Val-Leu-Trp

Biochem/physiol Actions

Selective endothelin A (ETA) antagonist

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ihara et al.
Life sciences, 50(4), 247-255 (1992-01-01)
We describe novel potent endothelin (ET) antagonists that are highly potent and selective for the ETA receptor (selective to ET-1). Of the synthetic analogs based on ETA antagonist BE-18257A isolated from Streptomyces misakiensis (IC50 value for ETA receptor on porcine
Agnes Boltz et al.
American journal of physiology. Heart and circulatory physiology, 304(1), H170-H174 (2012-10-30)
Endothelin-1 (ET-1) is an important regulator of vascular tone in the eye. It appears to play a role in ocular disease because of its strong vasoconstrictor action, its role in intraocular pressure homeostasis, and its neurotoxic potential. We have previously
K Wolff et al.
Acta obstetricia et gynecologica Scandinavica, 75(5), 432-438 (1996-05-01)
To compare the effect of endothelin on isolated resistance arteries from different vascular beds in normal and preeclamptic women before and after pretreatment with the ETA receptor antagonist BQ-123. Resistance arteries from myometrial and omental biopsies obtained at cesarean section
Jianjun Zhou et al.
Hypertension (Dallas, Tex. : 1979), 62(3), 599-607 (2013-07-03)
Preeclampsia is a life-threatening pregnancy disorder. However, its pathogenesis remains unclear. We tested the hypothesis that gestational hypoxia induces preeclampsia-like symptoms via heightened endothelin-1 (ET-1) signaling. Time-dated pregnant and nonpregnant rats were divided into normoxic and hypoxic (10.5% O2 from
Giuseppe Faraco et al.
Hypertension (Dallas, Tex. : 1979), 62(4), 759-766 (2013-08-21)
Endothelin-1 (ET1) is a potent vasoconstrictor peptide implicated in the cerebrovascular alterations occurring in stroke, subarachnoid hemorrhage, and brain trauma. Brain or circulating levels of ET1 are elevated in these conditions and in risk factors for cerebrovascular diseases. Most studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service