BM991.8613.02
B Medical U501
Ultra Low Temperature Freezer
Synonym(s):
B Medical, Ultra Low Temperature Freezer
About This Item
Recommended Products
AC/DC input
220 V / 240 V, US 3-pin plug
material
stainless steel device (Coated Steel Ext Stainless Steel Int)
feature
CE compliant
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
B Medical Systems
parameter
-40--86 °C temp. range
H × W × D
78.27 in. × 33.27 in. × 40.91 in.
capacity
21.118 ft3
weight
608.48 lb
shipped in
ambient
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
- Premium - Upright, Ultra Low Temperature Freeze
- 3 ST Shelves
- 4 Casters with integrated feet
- 7" touchscreen Display
- Alarm System w/ Battery Back-Up
- USB Data Port
- Door Mounted Control Panel
- Insulated Inner Doors
- Proximity Card Access Control
- Electronic Door Access
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service