Skip to Content
Merck
All Photos(1)

Key Documents

BM0033

Sigma-Aldrich

BMS-665053

≥98% (HPLC)

Synonym(s):

(S)-5-Chloro-1-(1-cyclopropylethyl)-3-(2,6-dichloro-4-(difluoromethoxy)phenylamino)-pyrazin-2(1H)-one, 5-Chloro-1-[(1S)-1-cyclopropylethyl]-3-[[2,6-dichloro-4-(difluoromethoxy)phenyl]amino]-2(1H)-pyrazinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14Cl3F2N3O2
CAS Number:
Molecular Weight:
424.66
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +16 to +21°, c = 0.4 in chloroform-d

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=CC(OC(F)F)=CC(Cl)=C1NC2=NC(Cl)=CN([C@@H](C)C3CC3)C2=O

Biochem/physiol Actions

BMS-665053 is a pyrazinone-containing antagonist that targets corticotropin-releasing factor/hormone (CRF or CRH) receptor 1 (CRF1, CRF-R1, CRFR-1, CRH-R1, CRHR-1) with high affinity (IC50 </= 1.0 nM against 150 pM ovine CRF for binding human and rat CRF-R1), potency (IC50 = 4.9 nM against 1 nM CRF-stimulated cAMP production in human Y-79 retinoblastoma cells), and selectivity, displaying no affinity toward CRF-R2/CRF2 (IC50 >10 μM against 150 pM ovine CRF in binding assay). BMS-665053 exhibits anxiolytic efficacy in a defensive withdrawal anxiety test in rats in vivo (10 mg/kg p.o.) with good oral bioavailability (F = 52%).
BMS-665053 is a pyrazinone-containing antagonist that targets corticotropin-releasing factor/hormone (CRF or CRH) receptor 1 (CRF1, CRF-R1, CRFR-1, CRH-R1, CRHR-1).

Features and Benefits

BMS-665053 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard A Hartz et al.
Journal of medicinal chemistry, 52(14), 4161-4172 (2009-06-26)
A series of pyrazinone-based heterocycles was identified as potent and orally active corticotropin-releasing factor-1 (CRF(1)) receptor antagonists. Selected compounds proved efficacious in an anxiety model in rats; however, pharmacokinetic properties were not optimal. In this article, we describe an in
Xiaoliang Zhuo et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(1), 5-15 (2009-10-17)
(S)-5-Chloro-1-(1-cyclopropylethyl)-3-(2,6-dichloro-4-(trifluoromethyl)phenylamino)pyrazin-2(1H)-one (BMS-665053), a pyrazinone-containing compound, is a potent and selective antagonist of corticotropin-releasing factor receptor-1 (CRF-R1) that showed efficacy in the defensive withdrawal model for anxiety in rats, suggesting its use as a potential treatment for anxiety and depression. In
William D Schmitz et al.
Bioorganic & medicinal chemistry letters, 20(12), 3579-3583 (2010-05-21)
A series of 5-arylamino-1,2,4-triazin-6(1H)-ones was synthesized and evaluated as antagonists at the corticotropin releasing factor receptor. Formation of CYP-mediated oxidative reactive metabolites previously observed in a related N(3)-phenylpyrazinone structure was minimized by incorporation of the additional ring nitrogen found in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service