Skip to Content
Merck
All Photos(1)

Key Documents

BM0019

Sigma-Aldrich

BMS-777607

≥98% (HPLC)

Synonym(s):

BMS-817378, N-(4-(2-Amino-3-chloropyridin-4-yloxy)-3-fluorophenyl)-4-ethoxy-1-(4-fluorophenyl)-2-oxo-1,2-dihydropyridine-3-carboxamide, N-[4-[(2-Amino-3-chloro-4-pyridinyl)oxy]-3-fluorophenyl]-4-ethoxy-1-(4-fluorophenyl)-1,2-dihydro-2-oxo-3-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19ClF2N4O4
CAS Number:
Molecular Weight:
512.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=C(N)N=CC=C1OC2=CC=C(NC(C3=C(OCC)C=CN(C4=CC=C(F)C=C4)C3=O)=O)C=C2F

InChI

1S/C25H19ClF2N4O4/c1-2-35-19-10-12-32(16-6-3-14(27)4-7-16)25(34)21(19)24(33)31-15-5-8-18(17(28)13-15)36-20-9-11-30-23(29)22(20)26/h3-13H,2H2,1H3,(H2,29,30)(H,31,33)

InChI key

VNBRGSXVFBYQNN-UHFFFAOYSA-N

Gene Information

human ... MET(4233)

Biochem/physiol Actions

BMS-777607 is an ATP-competitive inhibitor of the Met kinase superfamily with IC50 values of 1.8, 3.9, 4.3, and 1.1 nM, respectively, for Ron, Met, Tyro-3, and AxI kinases. BMS-777607 has been studied in a variety of cancer models and has shown anti-cancer activity, causing increased apoptosis, and decreased proliferation and migration, although it has also been shown to induce polyploidy, which may lead to chemoresistance.
BMS-777607 is an ATP-competitive inhibitor of the Met kinase superfamily.
RON kinase inhibitor (RONi, BMS-777607) decreases recepteur d′origine nantais (RON) signaling. This Met kinase inhibitor represses prostate cancer metastatic phenotype in vitro, induced by hepatocyte growth factor.

Features and Benefits

BMS-777607 is available through a partnership with Bristol-Myers Squibb (BMS). To learn more and view other BMS compounds, visit sigma.com/BMS.

Legal Information

Sold for research purposes only under agreement from BMS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RON kinase inhibition reduces renal endothelial injury in sickle cell disease mice.
Khaibullina A, et al.
Haematologica, 103(5), 787?798-787?798 (2018)
Cytokines: Advances in Research and Application: 2011 Edition, 548-548 (2012)
Dana Bohan et al.
PLoS pathogens, 17(11), e1009743-e1009743 (2021-11-20)
Phosphatidylserine (PS) receptors enhance infection of many enveloped viruses through virion-associated PS binding that is termed apoptotic mimicry. Here we show that this broadly shared uptake mechanism is utilized by SARS-CoV-2 in cells that express low surface levels of ACE2.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service