Skip to Content
Merck
All Photos(1)

Key Documents

B9553

Sigma-Aldrich

Monoclonal Anti-Bone Morphogenetic Protein 2 antibody produced in mouse

clone 65529.111, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-BMP-2

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
R$6,380.00

R$6,380.00


Estimated to ship onMay 30, 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1489


Select a Size

Change View
0.5 MG
R$6,380.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

R$6,380.00


Estimated to ship onMay 30, 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1489

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

65529.111, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

western blot: 1-2 μg/mL

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BMP2(650)

General description

Bone morphogenetic proteins (BMPs) are the members of the TGF-β superfamily and are identified as the critical factors for formation of bone and cartilage. Apart from this BMPs have immune regulatory functions. The cellular effects of BMPs are mediated by a heteromeric complex composed of type I and type II receptors. Among the BMPs, BMP-2 has bone and cartilage inducing abilities. The other important functions of BMP-2 are in heart formation. BMP-2 signaling also involves active cross talk with major pathways such as Smad, MAPK, PI3K and NF-κ B.. Monoclonal Anti-BMP-2 recognizes recombinant BMP-2. It also detects BMP-4 (25% cross reactivity).

Specificity

The antibody recognizes recombinant human BMP-2. It shows approximately 25% cross-reactivity with recombinant human BMP-4.

Immunogen

recombinant human BMP-2.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Monoclonal Anti-BMP-2 antibody may be used in immunoblotting at a working antibody concentration of 1-2 μg/ml.

Physical form

Lyophilizd from a 0.2 μm filtered solution in PBS, pH 7.4, with 5% trehalose.

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suzanne N Lissenberg-Thunnissen et al.
International orthopaedics, 35(9), 1271-1280 (2011-06-24)
This review evaluates the application of bone morphogenetic proteins (BMPs) in delayed bone repair, aiming at a broad audience from clinicians to scientists. Next to an overview of the role of the different BMPs, their antagonists and their current applications
Julia K Pester et al.
GMS Interdisciplinary plastic and reconstructive surgery DGPW, 2, Doc09-Doc09 (2013-01-01)
To determine bone morphogenetic protein (BMP)-2 protein and Aggrecan in osteoarthritic and healthy cartilage with special regard to localization and degree of cartilage damage 95 samples representing osteoarthritic cartilage and 17 samples out of normal cartilage were graded histological by
P C Bessa et al.
Journal of tissue engineering and regenerative medicine, 2(1), 1-13 (2008-02-23)
Discovered in 1965, bone morphogenetic proteins (BMPs) are a group of cytokines from the transforming growth factor-beta (TGFbeta) superfamily with significant roles in bone and cartilage formation. BMPs are used as powerful osteoinductive components of diverse tissue-engineering products for the
Flora Gilde et al.
Acta biomaterialia, 46, 55-67 (2016-09-17)
Surface coatings delivering BMP are a promising approach to render biomaterials osteoinductive. In contrast to soluble BMPs which can interact with their receptors at the dorsal side of the cell, BMPs presented as an insoluble cue physically bound to a
Kohei Miyazono et al.
Cytokine & growth factor reviews, 16(3), 251-263 (2005-05-06)
Bone morphogenetic proteins (BMPs), members of the transforming growth factor-beta (TGF-beta) superfamily, bind to two different serine/threonine kinase receptors, and mediate their signals through Smad-dependent and Smad-independent pathways. Receptor regulated-Smad (R-Smad) proteins specific for the BMP pathways interact with various

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service