Skip to Content
Merck
All Photos(1)

Key Documents

B8254

Sigma-Aldrich

Boc-Lys(Z)-OH

≥98%

Synonym(s):

Nα-Boc-Nε-Cbz-L-lysine, Nα-Boc-Nε-Z-L-lysine, Nε-Z-Nα-Boc-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28N2O6
CAS Number:
Molecular Weight:
380.44
Beilstein:
1917222
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

product name

Boc-Lys(Z)-OH,

Assay

≥98%

form

powder

color

white

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C19H28N2O6/c1-19(2,3)27-18(25)21-15(16(22)23)11-7-8-12-20-17(24)26-13-14-9-5-4-6-10-14/h4-6,9-10,15H,7-8,11-13H2,1-3H3,(H,20,24)(H,21,25)(H,22,23)/t15-/m0/s1

InChI key

BDHUTRNYBGWPBL-HNNXBMFYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nε-Z-Nα-Boc-L-lysine is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make peptides containing Nepsilon protected lysyl side chains.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service