B7138
Bispyrazolone
≥98%, crystalline
Synonym(s):
3,3′-Dimethyl-1,1′-diphenyl-(4,4′-bi-2-pyrazoline)-5,5′-dione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H18N4O2
CAS Number:
Molecular Weight:
346.38
Beilstein:
572787
EC Number:
MDL number:
UNSPSC Code:
12000000
Recommended Products
Assay
≥98%
form
crystalline
color
tan
suitability
suitable for determination of cyanide
SMILES string
CC1=NN(c2ccccc2)C(=O)C1C3C(C)=NN(c4ccccc4)C3=O
InChI
1S/C20H18N4O2/c1-13-17(19(25)23(21-13)15-9-5-3-6-10-15)18-14(2)22-24(20(18)26)16-11-7-4-8-12-16/h3-12,17-18H,1-2H3
InChI key
FORCWSNQDMPPOC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Epstein, J.
Analytical Chemistry, 19, 272-272 (1947)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service