Skip to Content
Merck
All Photos(1)

Documents

B6388

Sigma-Aldrich

Boc-Phe-Ser-Arg-7-amido-4-methylcoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H43N7O8
CAS Number:
Molecular Weight:
665.74
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥97% (TLC)

form

powder

solubility

DMSO: 20 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(NC(=O)C(CCCNC(N)=N)NC(=O)C(CO)NC(=O)C(Cc3ccccc3)NC(=O)OC(C)(C)C)ccc12

InChI

1S/C33H43N7O8/c1-19-15-27(42)47-26-17-21(12-13-22(19)26)37-28(43)23(11-8-14-36-31(34)35)38-30(45)25(18-41)39-29(44)24(16-20-9-6-5-7-10-20)40-32(46)48-33(2,3)4/h5-7,9-10,12-13,15,17,23-25,41H,8,11,14,16,18H2,1-4H3,(H,37,43)(H,38,45)(H,39,44)(H,40,46)(H4,34,35,36)

InChI key

JLKJMNJZJBEYLQ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Fukusen et al.
Journal of biochemistry, 119(4), 633-638 (1996-04-01)
Novel trypsin-like serine proteases (mouse trypsin-type serine proteases 1 and 2 [MTSP-1 and -2]) were purified to homogeneity from mouse spleen. Each protease consisted of a single polypeptide with a molecular mass of about 29 kDa, as determined by sodium
Y Suzuki et al.
Archives of dermatological research, 285(6), 372-377 (1993-01-01)
In order to identify the endogenous protease associated with stratum corneum (SC) desquamation, we examined properties of proteases in the stratum corneum of normal human skin. SC were obtained by tape stripping, washed in toluene and then dried. The proteolytic
T Kambara et al.
Experientia, 42(2), 155-157 (1986-02-15)
Proteolytic enzymes of the guinea pig peritoneal exudate macrophages were investigated using synthetic fluorogenic peptide substrates. Among several enzymes, t-butyloxycarbonyl-phenylalanyl-seryl-arginine 4-methylcoumaryl-7-amide cleaving enzymes had the highest activity, and the activity in exudate macrophages was about 3 times stronger than that
L Fiorucci et al.
FEBS letters, 408(1), 85-88 (1997-05-12)
Tryptases are oligomeric enzymes localized in the secretory granules of mast cells. Their role(s) in vivo has yet to be clarified and the lack of powerful and specific inhibitors has hampered the comprehension of the biological functions of these enzymes.
K Nakayama et al.
The Journal of biological chemistry, 270(40), 23619-23626 (1995-10-06)
Arginine-specific cysteine proteinase (Arg-gingipain; formerly, argingipain) is one of the major extracellular proteinases produced by the oral anaerobic bacterium Porphyromonas gingivalis. To determine whether Arg-gingipain is important for periodontopathogenicity of the organism, Arg-gingipain-deficient mutants were constructed via gene disruption by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service