B6029
Nα-Adamantaneacetyl-D-Arg-[Hyp3, Thi5,8, D-Phe7]-Bradykinin
≥97% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C68H99N19O14S2
CAS Number:
Molecular Weight:
1470.76
MDL number:
UNSPSC Code:
12352200
Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
NC(=N)NCCC[C@H](NC(=O)[C@H](Cc1cccs1)NC(=O)[C@@H](Cc2ccccc2)NC(=O)[C@H](CO)NC(=O)[C@H](Cc3cccs3)NC(=O)CNC(=O)[C@@H]4C[C@@H](O)CN4C(=O)[C@@H]5CCCN5C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](CCCNC(N)=N)NC(=O)CC67CC8CC(CC(C8)C6)C7)C(O)=O
Gene Information
human ... BDKRB2(624)
mouse ... BDKRB2(12062)
rat ... BDKRB2(25245)
Amino Acid Sequence
Nα-Adamantaneacetyl-D-Arg-Arg-Pro-Hydroxy-Pro-Gly-β-(2-Thienyl)Ala-Ser-D-Phe-β-(2-Thienyl)Ala-Arg
Biochem/physiol Actions
A highly potent antagonist at bradykinin B2 receptors with no agonist activity.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
B Lammek et al.
Peptides, 11(5), 1041-1043 (1990-09-01)
We report that the acylation of the N-terminus of [D-Arg0,Hyp3,Thi5,8,D-Phe7]bradykinin, one of the most potent bradykinin antagonists described to date, with 1-adamantane-acetic acid, results in an analogue with more than ten times enhanced potency. The new analogue does not increase
J A García-Sáinz et al.
Biochemistry and molecular biology international, 47(6), 927-933 (1999-07-20)
In C9 rat liver cells bradykinin and kallidin increased (approximately 2-fold) the intracellular concentration of calcium, but the B1 agonist, des-Arg9-bradykinin did not. The effect of bradykinin was inhibited by the B2 antagonists, Hoe 140 and N-alpha-adamantaneacetyl-D-Arg-[Hyp3, Thi5,8, D-Phe7]-bradykinin, but
Man Zhang et al.
American journal of physiology. Cell physiology, 296(6), C1364-C1372 (2009-04-03)
Bradykinin (BK) is an endogenous peptide with diverse biological actions and is considered to be an important mediator of the inflammatory response in both the peripheral and the central nervous systems. BK has attracted recent interest as a potential mediator
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service