Skip to Content
Merck
All Photos(1)

Documents

B0632

Sigma-Aldrich

Bradykinin potentiator C

≥97% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H77N11O13
CAS Number:
Molecular Weight:
1052.22
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥97% (TLC)

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C1CCCN1C(=O)C2CCCN2C(=O)CNC(=O)C3CCCN3C(=O)C4CCCN4C(=O)C(CC(C)C)NC(=O)CNC(=O)C5CCC(=O)N5)C(=O)N6CCCC6C(=O)N7CCCC7C(O)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S I Shimuta et al.
European journal of pharmacology, 70(4), 551-558 (1981-04-09)
The sensitivity of the guinea pig isolated ileum to bradykinin, but not to other agonists, was increased ca. 2-fold during the 3-4 h following mounting of the preparation under 1 g load. Concomitantly, a decrease was observed in the bradykinin
P Boschcov et al.
British journal of pharmacology, 83(2), 591-600 (1984-10-01)
Low doses of bradykinin (below 10 nM), as well as of K+ (below 10 mM) induced relaxation, whereas higher doses caused contraction of the rat duodenum. The relaxant responses induced by bradykinin and K+ were not affected by ouabain (1
Bradykinin-potentiating peptides from the venom of Agkistrodon halys blomhoffi. Isolation of five bradykinin potentiators and the amino acid sequences of two of them, potentiators B and C.
H Kato et al.
Biochemistry, 10(6), 972-980 (1971-03-16)
M Tominaga et al.
Journal of medicinal chemistry, 18(2), 130-133 (1975-02-01)
In a study of the relationship between structure and activity of bradykinin potentiating peptides (BPP), six analogs and homologs of peptides occurring in the venoms of Bothrops jararaca and Agkistrodon halys blomhoffii were synthesized and assayed in the isolated guinea

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service