Skip to Content
Merck
All Photos(1)

Key Documents

B0435

Sigma-Aldrich

Bohemine

≥95% (HPLC), powder

Synonym(s):

2-(3-Hydroxypropylamino)-6-benzylamino-9-isopropylpurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N6O
CAS Number:
Molecular Weight:
340.42
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥95% (HPLC)

form

powder

solubility

chloroform: soluble 50 mg/mL

storage temp.

−20°C

SMILES string

CC(C)n1cnc2c(NCc3ccccc3)nc(NCCCO)nc12

InChI

1S/C18H24N6O/c1-13(2)24-12-21-15-16(20-11-14-7-4-3-5-8-14)22-18(23-17(15)24)19-9-6-10-25/h3-5,7-8,12-13,25H,6,9-11H2,1-2H3,(H2,19,20,22,23)

InChI key

OPQGFIAVPSXOBO-UHFFFAOYSA-N

Biochem/physiol Actions

Cdk inhibitor; tool for studying cell cycle and anti-cancer activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service