Skip to Content
Merck
All Photos(2)

Key Documents

AV54594

Sigma-Aldrich

Anti-ARCN1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Archain 1, Anti-COPD

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onMay 28, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

dog, mouse, bovine, human, guinea pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARCN1(372)

Immunogen

Synthetic peptide directed towards the middle region of human ARCN1

Application

Anti-ARCN1 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

ARCN1 gene encodes an intracellular protein, which is the δ subunit of coat protein I (COPI) complex and is localized on chromosome 11 at 11q23.3. It is a 57kD protein consisting of C-terminal domain (CTD) and an N-­terminal longin domain. The encoded protein facilitates the retrograde transport of proteins and lipids from the cis-Golgi network to the endoplasmic reticulum and intra-Golgi membranes. Single nucleotide polymorphism in ARCN1 gene increases the risk of Glioma.

Sequence

Synthetic peptide located within the following region: GGLQNMELHGMIMLRISDDKYGRIRLHVENEDKKGVQLQTHPNVDKKLFT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Deng et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 68(Pt 7), 829-831 (2012-07-04)
Coat protein I (COPI) is a protein complex composed of seven subunits that mediates retrograde transport of proteins and lipids from the cis-Golgi network to the endoplasmic reticulum and intra-Golgi membranes. The medium-sized δ subunit of COPI (δ-COP) is a
Hongyan Chen et al.
PloS one, 7(12), e52864-e52864 (2013-01-10)
A single nucleotide polymorphism (SNP) at locus 11q23.3 (rs498872) in the near 5'-UTR of the PHLDB1 gene was recently implicated as a risk factor for gliomas in a genome-wide association study, and this involvement was confirmed in three additional studies.
Todd D Taylor et al.
Nature, 440(7083), 497-500 (2006-03-24)
Chromosome 11, although average in size, is one of the most gene- and disease-rich chromosomes in the human genome. Initial gene annotation indicates an average gene density of 11.6 genes per megabase, including 1,524 protein-coding genes, some of which were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service