Skip to Content
Merck
All Photos(1)

Key Documents

AV53656

Sigma-Aldrich

Anti-LGALS8 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Gal-8, Anti-Lectin, galactoside-binding, soluble, 8 (galectin 8), Anti-PCTA-1, Anti-PCTA1, Anti-Po66-CBP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onMay 21, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onMay 21, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

dog, human, rabbit, bovine, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LGALS8(3964)

General description

LGALS8 (lectin, galactoside-binding, soluble, 8) encodes a protein that belongs to galectin family and is predominantly expressed in tumoral tissues and may be involved in integrin-like cell interactions.

The previously assigned protein identifier Q9BXC8 has been merged into O00214. Full details can be found on the UniProt database.

Immunogen

Synthetic peptide directed towards the C terminal region of human LGALS8

Application

Anti-LGALS8 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

Gal-8 activates Rho signaling in TM cells and hence facilitates the regulation of cytoskeletal rearrangement in trabecular meshwork cells. Galectin-8 interacts with integrin and modulates its interaction with the extracellular matrix and hence inhibits cell adhesion and induces apoptosis. Additionally, it also has a role in modulating cell adhesion and cell growth.

Sequence

Synthetic peptide located within the following region: FPFSPGMYFEMIIYCDVREFKVAVNGVHSLEYKHRFKELSSIDTLEINGD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiri Diskin et al.
PloS one, 7(9), e44400-e44400 (2012-09-14)
The trabecular meshwork (TM) cell-matrix interactions and factors that influence Rho signaling in TM cells are thought to play a pivotal role in the regulation of aqueous outflow. The current study was designed to evaluate the role of a carbohydrate-binding
Yehiel Zick et al.
Glycoconjugate journal, 19(7-9), 517-526 (2004-02-06)
Galectin-8 belongs to the family of tandem-repeat type galectins. It consists as several isoforms, each made of two domains of approximately 140 amino-acids, both having a carbohydrate recognition domain (CRD). These domains are joined by a 'link peptide' of variable
Y R Hadari et al.
Journal of cell science, 113 ( Pt 13), 2385-2397 (2000-06-15)
The interaction of cells with the extracellular matrix regulates cell adhesion, motility, growth, survival and differentiation through integrin-mediated signal transduction. Here we demonstrate that galectin-8, a secreted mammalian (beta)-galactoside binding protein, inhibits adhesion of human carcinoma (1299) cells to plates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service