Skip to Content
Merck
All Photos(4)

Key Documents

AV51692

Sigma-Aldrich

Anti-MICALL1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp686M2226, Anti-FLJ45921, Anti-KIAA1668, Anti-MICAL-L1, Anti-MICAL-like 1, Anti-MIRAB13

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R$3,345.00

R$3,345.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
100 μL
R$3,345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,345.00


Estimated to ship onMay 28, 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

93 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MICALL1(85377)

Immunogen

Synthetic peptide directed towards the N terminal region of human MICALL1

Application

Anti-MICALL1 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5 μg/ml.

Biochem/physiol Actions

Molecule Interacting with CasL (MICAL)-like1 (MICALL1) is an endocytic regulatory protein that interacts with GTP-binding proteins such as Rab35. This interaction enhances the localization of MICALL1 to tubular recycling endosomes. MICALL1 also interacts with Rab13 and mediates the endocytosis of epidermal growth factor and regulates assembly of tight junction in the epithelial cells.

Sequence

Synthetic peptide located within the following region: ENGPEEGTFVCAEHCARLGPGTRSGTRPGPFSQPKQQHQQQLAEDAKDVP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliati Rahajeng et al.
Traffic (Copenhagen, Denmark), 13(1), 82-93 (2011-09-29)
Endocytosis is a conserved process across species in which cell surface receptors and lipids are internalized from the plasma membrane. Once internalized, receptors can either be degraded or be recycled back to the plasma membrane. A variety of small GTP-binding
Anne-Marie Marzesco et al.
Molecular biology of the cell, 13(6), 1819-1831 (2002-06-12)
Junctional complexes such as tight junctions (TJ) and adherens junctions are required for maintaining cell surface asymmetry and polarized transport in epithelial cells. We have shown that Rab13 is recruited to junctional complexes from a cytosolic pool after cell-cell contact
Nancy Abou-Zeid et al.
Molecular biology of the cell, 22(18), 3431-3441 (2011-07-29)
Small GTPase Rabs are required for membrane protein sorting/delivery to precise membrane domains. Rab13 regulates epithelial tight junction assembly and polarized membrane transport. Here we report that Molecule Interacting with CasL (MICAL)-like1 (MICAL-L1) interacts with GTP-Rab13 and shares a similar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service